Rat sL-Selectin ELISA kit
Fournisseur: Biorbyt
ORB106606-96
BIRBORB106606-96EA 0 EUR InStock
BIRBORB106606-96
Rat sL-Selectin ELISA kit
Tests Tests ELISA
Rat sL-Selectin ELISA Kit for the quantitative measurement of rat sL-selectin.
L-selectin, also known as CD62L, is a cell adhesion molecule found on leukocytes. It belongs to the selectin family of proteins, which recognise sialylated carbohydrate groups. It is cleaved by ADAM17.SELL. L-selectin is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains – one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins. L-selectin acts as a "homing receptor" for leukocytes to enter secondary lymphoid tissues via high endothelial venules. Ligands present on endothelial cells will bind to leukocyte expressing L-selectin, slowing leukocyte trafficking through the blood, and facilitating entry into a secondary lymphoid organ at that point.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité