Human FGF4 ELISA kit
Fournisseur: Biorbyt
ORB138053-96
BIRBORB138053-96EA 0 EUR InStock
BIRBORB138053-96
Human FGF4 ELISA kit
Tests Tests ELISA
Human FGF4 ELISA Kit for the quantitative detection of human fibroblast growth factor 4 (FGF-4) in cell culture supernates, serum and plasma (heparin, EDTA).
FGF-4 is a protein that in humans is encoded by the FGF4 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumour growth and invasion.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité