Human TIMP-4 ELISA kit
Fournisseur: Biorbyt
ORB138055-96
BIRBORB138055-96EA 0 EUR InStock
BIRBORB138055-96
Human TIMP-4 ELISA kit
Tests Tests ELISA
Human TIMP-4 ELISA Kit for the quantitative detection of human Tissue inhibitor of metalloproteinases 4 (TIMP-4) in cell culture supernates, serum, plasma (heparin, EDTA) and human milk.
Tissue inhibitor of metalloproteinases 4, also called Metalloproteinase inhibitor 4, is an enzyme that in humans is encoded by the TIMP4 gene. This gene belongs to the TIMP gene family. The proteins encoded by this gene family are inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix. This gene is mapped to 3p25.2. The secreted, netrin domain-containing protein encoded by this gene is involved in regulation of platelet aggregation and recruitment and may play role in hormonal regulation and endometrial tissue remodeling.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité