Human angiopoietin-1 ELISA kit
Fournisseur: Biorbyt
ORB138056-96
BIRBORB138056-96EA 0 EUR InStock
BIRBORB138056-96
Human angiopoietin-1 ELISA kit
Tests Tests ELISA
Human Angiopoietin-1 ELISA Kit is used for quantitative detection of human Angiopoietin-1 in cell culture supernates, serum, plasma (heparin, EDTA), saliva and human milk.
Angiopoietin 1, also called ANG1 is a type of angiopoietin and is encoded by the gene ANGPT1. Angiopoietins are proteins with important roles in vascular development and angiogenesis. The protein contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité