Human AXL ELISA kit
Fournisseur: Biorbyt
ORB138059-96
BIRBORB138059-96EA 0 EUR InStock
BIRBORB138059-96
Human AXL ELISA kit
Tests Tests ELISA
Human AXL ELISA Kit is used for quantitative detection of human AXL in cell culture supernates, serum and plasma (heparin, EDTA).
Tyrosine-protein kinase receptor UFO is an enzyme that in humans is encoded by the AXL gene. The protein encoded by this gene is a member of the receptor tyrosine kinase subfamily.
Axl is a chronic myelogenous leukemia-associated oncogene and also associated with colon cancer and melanoma. It is in close vicinity to the BCL3 oncogene, which is at 19q13.1-q13.2. The AXL gene is evolutionarily conserved between vertebrate species. This gene has two different alternatively spliced transcript variants.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité