Mouse TNFRSF7 ELISA kit
Fournisseur: Biorbyt
ORB138064-96
BIRBORB138064-96EA 0 EUR InStock
BIRBORB138064-96
Mouse TNFRSF7 ELISA kit
Tests Tests ELISA
Mouse TNFRSF7 ELISA Kit for the quantitative detection of mouse TNFRSF7 (CD27) in cell culture supernates and serum.
CD27 antigen, also called CD27 or TNFRSF7 is a tumour necrosis factor receptor. This gene is mapped to 12p13.31. The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is required for generation and long-term maintenance of T cell immunity. It binds to ligand CD70, and plays a key role in regulating B-cell activation and immunoglobulin synthesis. This receptor transduces signals that lead to the activation of NF-kappaB and MAPK8/JNK. Adaptor proteins TRAF2 and TRAF5 have been shown to mediate the signaling process of this receptor. CD27-binding protein (SIVA), a proapoptotic protein, can bind to this receptor and is thought to play an important role in the apoptosis induced by this receptor.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité