Human FLRG ELISA kit
Fournisseur: Biorbyt
ORB138072-96
BIRBORB138072-96EA 0 EUR InStock
BIRBORB138072-96
Human FLRG ELISA kit
Tests Tests ELISA
Human FLRG/FSTL3 ELISA kit for the quantitative detection of human FLRG in cell culture supernates, serum, plasma (heparin, EDTA), urine and human milk.
FSTL3 (follistatin-like 3) also known as FLRG or follistatin-related gene, is a member of the follistatin-module protein family, which is composed of extracellular matrix-associated glycoproteins thought to act in a paracrine manner to bind morphogens or growth/differentiation factors and regulate their activity during development. The FSTL3 gene extends over 7 kbp and contains 5 exons. The FSTL3 gene was localised to chromosome 19p13. This gene involved in hematopoiesis and in differentiation of hemopoietic progenitor cells, increases hematopoietic cell adhesion to fibronectin and seems to contribute to the adhesion of hematopoietic precursor cells to the bone marrow stroma. It may have a role in leukemogenesis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité