Human PLAT ELISA kit
Fournisseur: Biorbyt
ORB138073-96
BIRBORB138073-96EA 0 EUR InStock
BIRBORB138073-96
Human PLAT ELISA kit
Tests Tests ELISA
Human PLAT ELISA kit for the quantitative detection of human plasminogen activator, tissue in cell culture supernates, serum, plasma (heparin, EDTA, citrate) and urine.
Plasminogen activator, tissue, also called PLAT or TPA is a serine protease. This gene encodes tissue-type plasminogen activator, a secreted serine protease which converts the proenzyme plasminogen to plasmin, a fibrinolytic enzyme. Tissue-type plasminogen activator is synthesized as a single chain which is cleaved by plasmin to a two chain disulfide linked protein. This gene was mapped to 8p11.21. This enzyme plays a role in cell migration and tissue remodeling. Increased enzymatic activity causes hyperfibrinolysis, which manifests as excessive bleeding; decreased activity leads to hypofibrinolysis which can result in thrombosis or embolism.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité