Human TSLP ELISA kit
Fournisseur: Biorbyt
ORB138077-96
BIRBORB138077-96EA 0 EUR InStock
BIRBORB138077-96
Human TSLP ELISA kit
Tests Tests ELISA
Human TSLP ELISA kit for the quantitative detection of human thymic stromal lymphopoietin (TSLP) in cell culture supernates, serum and plasma (heparin, EDTA).
TSLP is a protein belonging to the cytokine family. This gene is mapped to 5q22.1. It encodes a hemopoietic cytokine proposed to signal through a heterodimeric receptor complex composed of the thymic stromal lymphopoietin receptor and the IL-7R alpha chain. It mainly impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes and enhances the maturation of CD11c(+) dendritic cells. The protein promotes T helper type 2 (TH2) cell responses that are associated with immunity in various inflammatory diseases, including asthma, allergic inflammation and chronic obstructive pulmonary disease. The protein is therefore considered a potential therapeutic target for the treatment of such diseases.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité