Human PD1 ELISA kit
Fournisseur: Biorbyt
ORB138078-96
BIRBORB138078-96EA 0 EUR InStock
BIRBORB138078-96
Human PD1 ELISA kit
Tests Tests ELISA
Human PD1 ELISA kit for the quantitative detection of human programmed cell death protein 1 in cell culture supernates.
Programmed cell death protein 1, also called PD-1 or PDCD1 is a protein that in humans is encoded by the PDCD1 gene. This gene encodes a cell surface membrane protein of the immunoglobulin superfamily. By fluorescence in situ hybridisation, the PDCD1 gene is mapped to 2q37.3. This protein is expressed in pro-B-cells and is thought to play a role in their differentiation. In mice, expression of this gene is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure. These studies suggest that this gene product may also be important in T cell function and contribute to the prevention of autoimmune diseases.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité