Human ANGPTL4 ELISA kit
Fournisseur: Biorbyt
ORB138079-96
BIRBORB138079-96EA 0 EUR InStock
BIRBORB138079-96
Human ANGPTL4 ELISA kit
Tests Tests ELISA
Human ANGPTL4 ELISA kit is used for quantitative detection of human ANGPTL4 in cell culture supernates, serum and plasma (heparin, EDTA).
Angiopoietin-related protein 4 (ANGPTL4) is a protein that in humans is encoded by the ANGPTL4 gene. This gene is a member of the angiopoietin/angiopoietin-like gene family and encodes a glycosylated, secreted protein with a fibrinogen C-terminal domain.
This gene is induced under hypoxic conditions in endothelial cells and is the target of peroxisome proliferation activators. By radiation hybrid analysis, ANGPTL4 gene is mapped to 19p13.3. ANGPTL4 contributed to tumour growth and protected cells from anoikis, a form of programmed cell death induced when contact-dependent cells detach from the surrounding tissue matrix.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité