Human IGFBP6 ELISA kit
Fournisseur: Biorbyt
ORB138081-96
BIRBORB138081-96EA 0 EUR InStock
BIRBORB138081-96
Human IGFBP6 ELISA kit
Tests Tests ELISA
Human IGFBP6 ELISA kit for the quantitative detection of human insulin-like growth factor-binding protein 6 (IGFBP6) in cell culture supernates, serum and plasma (heparin, EDTA).
Insulin-like growth factor-binding protein 6 (IGFBP6) is a protein that in humans is encoded by the IGFBP6 gene. The IGFBP6 gene is mapped to chromosome 12. In all, it has contained 216 amino acid protein with a calculated molecular mass of 22847, The human keratinocyte cell line HaCat secretes IGFBP6 as an autocrine growth inhibitor. Recombinant IGFBP6 was also shown to inhibit growth of HaCat cells and other keratinocyte cell lines.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité