Human IL-17RC ELISA kit
Fournisseur: Biorbyt
ORB138088-96
BIRBORB138088-96EA 0 EUR InStock
BIRBORB138088-96
Human IL-17RC ELISA kit
Tests Tests ELISA
Human IL-17RC ELISA kit for the quantitative detection of human Interleukin-17 receptor C (IL-17RC) in cell culture supernates, serum and plasma (heparin, EDTA).
Interleukin-17 receptor C is a protein that in humans is encoded by the IL17RC gene. By genomic sequence analysis, IL17RC gene was mapped to chromosome 3p25.3-p24.1. This gene encodes a single-pass transmembrane protein that shares limited similarity with the interleukin-17 receptor. Multiple alternatively spliced transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only three have been determined to date.
The biologic activity of IL-17 is dependent on a complex composed of IL-17RA and IL-17RC, suggesting a new paradigm for understanding the interactions between the expanded family of IL-17 ligands and their receptors.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité