Human TNFSF14 ELISA kit
Fournisseur: Biorbyt
ORB138092-96
BIRBORB138092-96EA 0 EUR InStock
BIRBORB138092-96
Human TNFSF14 ELISA kit
Tests Tests ELISA
Human TNFSF14 ELISA kit for the quantitative detection of human tumour necrosis factor ligand superfamily member 14 (TNFSF14) in cell culture supernates, serum, plasma (heparin, EDTA) and saliva.
Tumour necrosis factor ligand superfamily member 14 is a protein that in humans is encoded by the TNFSF14 gene. TNFSF14 has also been designated as CD258, as well as LIGHT. It was mapped on chromosome 19p13.3. The protein encoded by this gene is a member of the tumor necrosis factor (TNF) ligand family. This protein may function as a costimulatory factor for the activation of lymphoid cells and as a deterrent to infection by herpesvirus. This protein has been shown to stimulate the proliferation of T cells, and trigger apoptosis of various tumor cells. This protein is also reported to prevent tumour necrosis factor alpha mediated apoptosis in primary hepatocyte. Two alternatively spliced transcript variant encoding distinct isoforms have been reported.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité