Human ALCAM ELISA kit
Fournisseur: Biorbyt
ORB138096-96
BIRBORB138096-96EA 0 EUR InStock
BIRBORB138096-96
Human ALCAM ELISA kit
Tests Tests ELISA
Human ALCAM ELISA Kit is used for the quantitative detection of human ALCAM in cell culture supernates, serum and plasma (heparin, EDTA).
CD166 antigen is a 100 to 105 kD type I transmembrane glycoprotein that is a member of the immunoglobulin superfamily of proteins. In humans it is encoded by the ALCAM gene. The gene was mapped to human chromosome 3q13.1-q13.2 by fluorescence in situ hybridisation of cDNA probes to metaphase chromosomes. It is also called CD166, MEMD, SC-1/DM-GRASP/BEN in the chicken, and KG-CAM in the rat.
It is expressed on activated T cells, activated monocytes, epithelial cells, fibroblasts, neurons, melanoma cells, and also in sweat and sebaceous glands. CD166 protein expression is reported to be upregulated in a cell line deriving from a metastasising melanoma.
CD166 plays an important role in mediating adhesion interactions between thymic epithelial cells and CD6+ cells during intrathymic T cell development. Recently, CD166 has also been used as a potential cancer stem cell marker.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité