Mouse MMP-12 ELISA kit
Fournisseur: Biorbyt
ORB138097-96
BIRBORB138097-96EA 0 EUR InStock
BIRBORB138097-96
Mouse MMP-12 ELISA kit
Tests Tests ELISA
Mouse MMP-12 ELISA kit for the quantitative detection of mouse Matrix metalloproteinase-12 in cell culture supernates, serum and plasma (heparin).
Matrix metalloproteinase-12 (MMP12), also known as MME or ME, is an enzyme that in humans is encoded by the MMP12 gene. The gene is part of a cluster of MMP genes which localise to chromosome 11q22.2. It is thought that the protein encoded by this gene is cleaved at both ends to yield the active enzyme, but this processing has not been fully described. The enzyme degrades soluble and insoluble elastin. It may play a role in aneurysm formation and studies in mice suggest a role in the development of emphysema. This gene may involved in tissue injury and remodeling.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité