Human ACE2 ELISA kit
Fournisseur: Biorbyt
ORB138098-96
BIRBORB138098-96EA 0 EUR InStock
BIRBORB138098-96
Human ACE2 ELISA kit
Tests Tests ELISA
Human ACE2 ELISA Kit is used for quantitative detection of human ACE2 in cell culture supernates, serum and plasma (heparin, EDTA).
Angiotensin I converting enzyme 2 (ACE 2) is a protein belongs to the angiotensin-converting enzyme family of dipeptidyl carboxydipeptidases and has considerable homology to human angiotensin 1 converting enzyme. By sequence similarity to a sequence in GenBank, this gene is mapped to Xp22.2. This secreted protein catalyses the cleavage of angiotensin I into angiotensin 1 - 9, and angiotensin II into the vasodilator angiotensin 1 to 7.
The organ- and cell-specific expression of this gene suggests that it may play a role in the regulation of cardiovascular and renal function, as well as fertility. In addition, the encoded protein is a functional receptor for the spike glycoprotein of the human coronaviruses SARS and HCoV-NL63.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité