Human granzyme B ELISA kit
Fournisseur: Biorbyt
ORB138107-96
BIRBORB138107-96EA 668 EUR InStock
BIRBORB138107-96
Human granzyme B ELISA kit
Tests Tests ELISA
Human granzyme B ELISA kit for the quantitative detection of human Granzyme B in cell culture supernates, serum and plasma (heparin, EDTA).
Granzyme B is a serine protease that in humans is encoded by the GZMB gene. Granzyme B is expressed by cytotoxic T lymphocytes (CTL) and natural killer (NK) cells. CTL and NK cells share the remarkable ability to recognise specific infected target cells. They are thought to protect their host by inducing apoptosis of cells that bear on their surface nonself antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein encoded by this gene is crucial for the rapid induction of target cell apoptosis by CTL in cell-mediated immune response.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité