Mouse FGF-21 ELISA kit
Fournisseur: Biorbyt
ORB54811-96
BIRBORB54811-96EA 0 EUR InStock
BIRBORB54811-96
Mouse FGF-21 ELISA kit
Tests Tests ELISA
Mouse FGF-21 ELISA kit is designed for the quantification of mouse fibroblast growth factor 21 (FGF-21) in serum, plasma and cell culture supernate samples.
Fibroblast growth factor 21 (FGF-21) is a novel protein that has been implicated in the regulation of lipid and glucose metabolism under fasting and ketotic conditions. In murine models, FGF-21 is predominantly expressed in liver, but it also expressed in adipose tissue and pancreatic cells. It also protects animals from diet-induced obesity when overexpressed in transgenic mice and lowers blood glucose and triglyceride levels when administered to diabetic rodents5.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité