Human lipocalin-2 ELISA kit
Fournisseur: Biorbyt
ORB54812-96
BIRBORB54812-96EA 0 EUR InStock
BIRBORB54812-96
Human lipocalin-2 ELISA kit
Tests Tests ELISA
Human lipocalin-2 ELISA kit for the quantification of human lipocalin-2 in serum, plasma, urine and cell culture supernate samples.
Lipocalin-2 (LCN2), also known as neutrophil gelatinase-associated lipocalin (NGAL), 24p3, or neutrophil lipocalin (NL), is a 25-kDa secretory glycoprotein. LCN2 has been implicated in a variety of cellular processes including the innate immune response, differentiation, tumorigenesis, and cell survival.
Information de livraison: Kit includes micro-titre strips (96 wells), wash buffer (10×40 ml), assay buffer (5×20 ml), detection antibody solution (100×0,12 ml), lyophilised human lipocalin-2 standard, substrate solution (12 ml), stop solution (12 ml).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité