Human FGF-19 ELISA kit
Fournisseur: Biorbyt
ORB54823-96
BIRBORB54823-96EA 0 EUR InStock
BIRBORB54823-96
Human FGF-19 ELISA kit
Tests Tests ELISA
Human FGF-19 ELISA kit for the quantitative determination of human fibroblast growth factor 19 (FGF-19).
FGF-19 is a member of a subfamily of FGFs that includes FGF-21 and FGF-23, each member functions as an important regular of nutrient metabolism. The primary source of endocrine FGF-19 is the ileum, bile acids release into the intestine after a meal to induce expression of FGF-19. Circulating FGF-19 plays an important role in maintaining proper bile acid homeostasis. Several pharmacologic studies in hyperglycaemic, obese animal models have shown that FGF-19 can improve metabolic rate and lower serum glucose and hepatic triglyceride and cholesterol levels. Like insulin, FGF-19 functions as postprandial hormone to govern hepatic protein synthesis, glycogen synthesis and gluconeogenesis, but does not stimulate lipogenesis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité