Human DKK1 ELISA kit
Fournisseur: Biorbyt
ORB96798-96
BIRBORB96798-96EA 0 EUR InStock
BIRBORB96798-96
Human DKK1 ELISA kit
Tests Tests ELISA
Human DKK1 ELISA Kit is a sandwich ELISA kit for the quantitative detection of human Dickkopf-related protein 1 (DKK1).
  • No detectable cross-reactivity with other relevant proteins
Dickkopf-related protein 1 is a protein that in humans is encoded by the DKK1 gene, which was mapped to chromosome 10q11.2. This gene encodes a protein that is a member of the dickkopf family. It is a secreted protein with two cysteine rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway. Elevated levels of DKK1 in bone marrow, plasma and peripheral blood is associated with the presence of osteolytic bone lesions in patients with multiple myeloma.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité