Human THBS1 ELISA kit
Fournisseur: Biorbyt
ORB96799-96
BIRBORB96799-96EA 0 EUR InStock
BIRBORB96799-96
Human THBS1 ELISA kit
Tests Tests ELISA
Human THBS1 ELISA Kit is a sandwich ELISA kit for the quantitative detection of human Thrombospondin 1.
  • No detectable cross-reactivity with other relevant proteins
Thrombospondin 1, also known as THBS1, is a protein that in humans in encoded by the THBS1 gene. Thrombospondin 1 is a subunit of a disulfide-linked homotrimeric protein. This protein is an adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. By in situ hybridization, the THBS1 gene was mapped to human 15q15 and the cognate gene to mouse chromosome 2 region F. In 1990, the THBS1 gene was localized to 15q11-qter by Southern analysis of human-rodent somatic cell hybrids. Thrombospondin I is a multimodular secreted protein that associates with the extracellular matrix and possesses a variety of biologic functions, including a potent antiangiogenic activity. Other thrombospondin genes include thrombospondins II, III, and IV.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité