Human IL-22 ELISA kit
Fournisseur: Biorbyt
ORB99233-96
BIRBORB99233-96EA 0 EUR InStock
BIRBORB99233-96
Human IL-22 ELISA kit
Tests Tests ELISA
Human IL-22 ELISA Kit for the quantitative detection of human Interleukin-22 in cell culture supernates, serum and plasma (heparin, EDTA).
  • No detectable cross-reactivity with other relevant proteins
Interleukin-22 (IL-22), also known as ILTIF, is protein that in humans is encoded by the IL22 gene. IL-22 a member of a group of cytokines called the IL-10 family or IL-10 superfamily a class of potent mediators of cellular inflammatory responses. Using FISH, the IL22 gene is mapped to chromosome 12q15, close to the IFNG and the herpesvirus saimiri-induced AK155 genes. IL-22 can contribute to immune disease through the stimulation of inflammatory responses, S100s and defensins. IL-22 also promotes hepatocyte survival in the liver and epithelial cells in the lung and gut similar to IL-10. In some contexts, the pro-inflammatory versus tissue-protective functions of IL-22 are regulated by the often co-expressed cytokine IL-17A.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité