Human bone marrow, unprocessed
Fournisseur: Charles River Laboratories Cell Solutions
BM008F-4
BM008F-4EA 0 EUR InStock
BM008F-4
Human bone marrow, unprocessed
Cellules
Bone marrow is a major source of stem cells, which are being used in cancer research, cell studies, transplantation studies, and other research.
  • Test negative for Hepatitis B and C, HIV 1/2, HTLV- I/II, HCV/HBV, WNV, Trypanasoma cruzi, and Syphilis
Bone marrow collected from healthy human volunteer donors under IRB approved informed consent, drawn from the posterior iliac crest using a syringe containing anticoagulant.
Attention: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité