Quantitative migroorganisms, BioBall® MultiShot 550 and MultiShot 10e8
56021 56014 56016 56145 56146 410106 56011 56018 56150 410386 56006 415159
BMER56021EA 114.1 EUR InStock
BMER56021 BMER56014 BMER56016 BMER56145 BMER56146 BMER410106 BMER56011 BMER56018 BMER56150 BMER410386 BMER56006 BMER415159
Quantitative migroorganisms, BioBall® MultiShot 550 and MultiShot 10e8
Tests microbiologiques Tests sur les micro-organismes
BioBall® contains a precise number of microorganisms in a water soluble ball delivering unprecedented accuracy for Quantitative Microbiological Quality Control. BioBall Multishot 550 – mean of between 500 and 600 cfu with a standard deviation of ≤10% of the mean. Designed for re-hydration into 1,1 ml of MultiShot 550 Re-Hydration Fluid to provide 10×100 µl doses of 50 cfu. Ideal for pharmaceutical microbiology QC where multiple positive controls in the 10 to 100 cfu range are needed.
BioBall MultiShot 10e8 – mean of between 0.7 and 1,5×108 cfu with a standard deviation of ≤20% of the mean. Designed for rehydration into 1,1 ml of BioBall Re-Hydration Fluid to provide 10×100 µl doses of 107. Pharmaceutical and cosmetic preparations often require antimicrobial activity to prevent microbial proliferation or to limit the effects of contamination. The Pharmacopoeias require testing these products for the efficacy of antimicrobial preservation (e.g. USP 51, EP 5.1.3, JP19). BioBall MultiShot 10E8 is ideal for this application and simplifies preparation and provides a precise inoculum.
  • Easy to use and requires no preparation or pre-incubation
  • Can be added directly to media or matrix samples
  • Accredited reference material under ISO Guide 34 standard
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité