Anti-KRT18 Mouse Monoclonal Antibody (Purified) [clone: DA7]
Fournisseur: Biotium
BNUB0198-500
BNUB0198-500EA 511 EUR InStock
BNUB0198-500
Anti-KRT18 Mouse Monoclonal Antibody (Purified) [clone: DA7]
Anticorps
This MAb reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It was reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité