Anti-TP53 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TRP/816]
Fournisseur: Biotium
BNUM0816-50
BNUM0816-50EA 417 EUR InStock
BNUM0816-50
Anti-TP53 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TRP/816]
Anticorps
Recognizes a 53 kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53 under denaturing and non-denaturing conditions. p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to MDM2, SV40 T antigen and human papilloma virus E6 protein. Positive nuclear staining with p53 antibody has been reported to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivityMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité