Anti-Ep-CAM Mouse Monoclonal Antibody (Purified, BSA-free) [clone: EGP40/837]
Fournisseur: Biotium
BNUM0837-50
BNUM0837-50EA 417 EUR InStock
BNUM0837-50
Anti-Ep-CAM Mouse Monoclonal Antibody (Purified, BSA-free) [clone: EGP40/837]
Anticorps
EGP40 is a 40-43 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas with the exception of adult squamous epithelium, hepatocytes and gastric epithelial cells. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma.
Type: Primary
Antigen: Ep-More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité