Anti-KRT76 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KRTH/1076]
Fournisseur: Biotium
BNUM1076-50
BNUM1076-50EA 417 EUR InStock
BNUM1076-50
Anti-KRT76 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KRTH/1076]
Anticorps
This MAb recognizes basic (Type II or HMW) cytokeratins, which include 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8). Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50 , 48, 46, 45, and 40 kDa. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Type: Primary
Antigen: KRT76
Clonality: Monoclonal
Clone: KRTH/1076
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité