Anti-NAPSA Mouse Monoclonal Antibody (Purified, BSA-free) [clone: NAPSA/1238]
Fournisseur: Biotium
BNUM1238-50
BNUM1238-50EA 417 EUR InStock
BNUM1238-50
Anti-NAPSA Mouse Monoclonal Antibody (Purified, BSA-free) [clone: NAPSA/1238]
Anticorps
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B.There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.
Type: Primary
Antigen:More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité