Sampling pumps, BOLA
Fournisseur: Bohlender
A124-16
BOHLA124-16EA 314 EUR InStock
BOHLA124-16
Sampling pumps, BOLA
Pompe Pompes pour conteneur/transfert
Sampling pump is made up of PP and PTFE.
  • Compact construction and easy handling
  • Quick and low-risk pumping of ultrapure liquids
  • Ergonomic and usable anywhere
  • Universal chemical resistance
A pull on the ball handle produces a slight vacuum in the sampling bottle. Due to this vacuum, the sample is sucked into the sampling bottle.
Glass bottles and plastic bottles with a GL 45 thread can be used as sampling bottles.
Applicable for pumping liquids from sources that are not easily accessible; also, suitable for liquids with a high viscosity like oils.
Information de livraison: Supplied with tubing outer Ø (6 mm), PTFE tubing (2 m).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité