Multiple distributors with ground joints, BOLA
Fournisseur: Bohlender
D620-08
BOHLD620-08EA 149 EUR InStock
BOHLD620-08
Multiple distributors with ground joints, BOLA
Adaptateurs Raccords rodés
PTFE, white. For bottles or reaction vessels with ground joint. For transferring liquids without contamination. For connection to a liquid source or a pressure or vacuum system.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
  • Vacuum: suitable
Completely made of PTFE. With ground joint size 29/32 and two GL-threaded necks. Integrated special nut for easy unlocking of the ground joint. For the connection of tubing or tube made of glass, plastic or metal by means of laboratory screw joints.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité