Desiccator cabinet, SICCO wall
Fournisseur: Bohlender
V1994-08
BOHLV1994-08EA 16890 EUR InStock
BOHLV1994-08
Desiccator cabinet, SICCO wall
Dessiccateurs Armoires de dessiccation
Transparent PPMA cabinet with reinforced aluminium frame, unit has a door with a foam rubber gasket and magnetic strip. Designed for storing or drying humidity sensitive products using silica gel. The controlled environment inside the desiccator is ideal for storing reference materials, retained samples, metallographic specimens, tobacco and DNA-samples.
  • Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Temperature resistance from −20 to +70 °C
  • Easy to move about as unit has four castors, two of them with brakes
Information de livraison: Unit has six compartments, each with its own door, removable stainless steel telescopic shelf, desiccant tray and hygrometer. Each chamber can accommodate maximum of 17 shelves.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité