Enzymatic Digestion Amylase
Fournisseur: BIO-OPTICA
04-140808
BOPT04-140808EA 29.2 EUR InStock
BOPT04-140808
Enzymatic Digestion Amylase
Enzymes
Product for the preparation of cyto-histological samples for optical microscopy.
  • Amylase from Bacillus subtilis in buffered solution pH 7,3: 30 ml
  • Hazardous preparation: Observe all state and local environmental regulations regarding waste disposal
  • Validity: One year
Removal of glycogen from tissue sections:
- Liver, paraffin embedded sections: The removal of glycogen is advisable in order to observe only epithelial neutral mucins. This method is recommended for liver biopsy.
- Muscle frozen section.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité