Anti-MS4A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-0080R-A350
BOSSBS-0080R-A350EA 893 EUR InStock
BOSSBS-0080R-A350
Anti-MS4A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
B lymphocytes marker CD20 is a non glycosylated protein with a molecular weight of 35 or 37 kDa depending on the degree of phosphorylation. Although not a member of the tetraspanin superfamily of cell surface receptors, it crosses the cell membrane four times. The CD20 antigen is present on human pre B lymphocytes and on B lymphocytes at all stages of maturation, except on plasma cells. Low level expression of the CD20 antigen has been detected on normal T lymphocytes. The CD20 molecule is involved in regulation of B cell differentiation, presumably via its reported function as a Ca++More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité