Anti-HSPA1A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: Bioss
BS-0126R-FITC
BOSSBS-0126R-FITCEA 909 EUR InStock
BOSSBS-0126R-FITC
Anti-HSPA1A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
In cooperation with other chaperones, Hsp7s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage. In case of rotavirus A infection, serves as a post-attachment receptor for the virus toMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité