Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-KLH Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Bioss
BS-0225M-HRP
BOSSBS-0225M-HRPEA 858 EUR InStock
BOSSBS-0225M-HRP
Anti-KLH Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Keyhole limpet hemocyanin is an extremely large, heterogeneous glycosylated protein consisting of subunits with a molecular weight of 350,000 and 390,000 in aggregates with molecular weights of 4,500,000-13,000,000. Each domain of a KLH subunit contains two copper atoms that together bind a single oxygen molecule (O2). When oxygen is bound to hemocyanin, the molecule takes on a distinctive transparent, opalescent blue color. The KLH protein is potently immunogenic yet safe in humans and is therefore highly prized as a vaccine carrier protein. The large and highly glycosylated KLH protein cannot be reproduced synthetically. It is available only as a purified biological product from the Keyhole Limpet Megathura crenulata.Keyhole limpet hemocyanin (KLH) is used extensively as a carrier protein in the production of for research, biotechnology and therapeutic applications. Haptens are substances with a low molecular weight such as peptides, small proteins and drug molecules that are generally not immunogenic and require the aid of a carrier protein to stimulate a response from the immune system in the form of antibody production.[2] KLH is the most widely employed carrier proteins for this purpose. KLH is an effective carrier protein for several reasons. Its large size and numerous epitopes generate a substantial immune response, and abundance of lysine residues for coupling haptens, allows a high hapten:carrier protein ratio increasing the likelihood of generating hapten-specific . In addition, because KLH is derived from the limpet, a gastropod, it is phylogenetically distant from mammalian proteins, thus reducing false positives in immunologically based research techniques in mammalian model organisms.KLH may also be a challenging molecule to work with because of its propensity to aggregate and precipitate. Aggregates remain immunogenic, but limit the ability to conjugate haptens and are difficult to manipulate in the laboratory.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Others

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité