Anti-C Peptide Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-0274R-A750
BOSSBS-0274R-A750EA 890 EUR InStock
BOSSBS-0274R-A750
Anti-C Peptide Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
C Peptide is part of the molecule of Proinsulin, that consists of three parts: C Peptide and two long strands of amino acids (called the alpha and beta chains) that later become linked together to form the insulin molecule. From every molecule of proinsulin, one molecule of insulin plus one molecule of C Peptide are produced. C peptide is released into the blood stream in equal amounts to insulin. A test of C peptide levels will show how much insulin the body is making. Insulin decreases blood glucose concentration. It increases cell permeability to monosaccharides, amino acids and fattyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité