Anti-PRKCA + PRKCB Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-0520R-CY5.5
BOSSBS-0520R-CY5.5EA 909 EUR InStock
BOSSBS-0520R-CY5.5
Anti-PRKCA + PRKCB Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
Mediates cAMP-dependent signaling triggered by receptor binding to GPCRs. PKA activation regulates diverse cellular processes such as cell proliferation, the cell cycle, and differentiation and regulation of microtubule dynamics, chromatin condensation and decondensation, nuclear envelope disassembly and reassembly, as well as regulation of intracellular transport mechanisms and ion flux. Regulates the abundance of compartmentalized pools of its regulatory subunits through phosphorylation of PJA2 which binds and ubiquitinates these subunits, leading to their subsequent proteolysis (ByMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité