Anti-PRKCA + PRKCB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-0520R-HRP
BOSSBS-0520R-HRPEA 909 EUR InStock
BOSSBS-0520R-HRP
Anti-PRKCA + PRKCB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Mediates cAMP-dependent signaling triggered by receptor binding to GPCRs. PKA activation regulates diverse cellular processes such as cell proliferation, the cell cycle, and differentiation and regulation of microtubule dynamics, chromatin condensation and decondensation, nuclear envelope disassembly and reassembly, as well as regulation of intracellular transport mechanisms and ion flux. Regulates the abundance of compartmentalized pools of its regulatory subunits through phosphorylation of PJA2 which binds and ubiquitinates these subunits, leading to their subsequent proteolysis (ByMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité