Anti-p19ARF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-0534R-A350
BOSSBS-0534R-A350EA 893 EUR InStock
BOSSBS-0534R-A350
Anti-p19ARF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
P19ARF Capable of inducing cell cycle arrest in G1 and G2 phases. Acts as a tumor suppressor. Binds to MDM2 and blocks its nucleocytoplasmic shuttling by sequestering it in the nucleolus. This inhibits the oncogenic action of MDM2 by blocking MDM2-induced degradation of p53 and enhancing p53-dependent transactivation and apoptosis. Also induces G2 arrest and apoptosis in a p53-independent manner by preventing the activation of cyclin B1/CDC2 complexes. Binds to BCL6 and down-regulates BCL6-induced transcriptional repression. Binds to E2F1 and MYC and blocks their transcriptional activatorMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité