Anti-Eph receptor B1 Tyr928 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-10138R-A750
BOSSBS-10138R-A750EA 917 EUR InStock
BOSSBS-10138R-A750
Anti-Eph receptor B1 Tyr928 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Receptor tyrosine kinase which binds promiscuously transmembrane ephrin-B family ligands residing on adjacent cells, leading to contact-dependent bidirectional Signalling into neighboring cells. The Signalling pathway downstream of the receptor is referred to as forward Signalling while the Signalling pathway downstream of the ephrin ligand is referred to as reverse Signalling. Cognate/functional ephrin ligands for this receptor include EFNB1, EFNB2 and EFNB3. During nervous system development, regulates retinal axon guidance redirecting ipsilaterally ventrotemporal retinal ganglion cellsMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité