Anti-CFI Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-10339R-A647
BOSSBS-10339R-A647EA 893 EUR InStock
BOSSBS-10339R-A647
Anti-CFI Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
The complement pathway is an important host defense system that contributes to both innate and acquired immunity. There are three pathways of complement activation: the classical pathway, lectin pathway and alternative pathway. Complement protein Factor I is a key serine protease that modulates the complement cascade by regulating the levels of C3 convertases. It circulates in plasma as a heavily N-glycosylated heterodimer made up of two disulfide linked chains, each carrying three N-linked oligosaccharide chains that may have both structural and functional roles in the interactions with theMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité