Anti-MYD88 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-1047R-CY3
BOSSBS-1047R-CY3EA 909 EUR InStock
BOSSBS-1047R-CY3
Anti-MYD88 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Adapter protein involved in the Toll-like receptor and IL-1 receptor signaling pathway in the innate immune response. Acts via IRAK1, IRAK2, IRF7 and TRAF6, leading to NF-kappa-B activation, cytokine secretion and the inflammatory response. Increases IL-8 transcription. Involved in IL-18-mediated signaling pathway. Isoform 2 is defective in its ability to induce IRAK phosphorylation and NF-kappa-B activation and can function as a negative regulator of activation by IL-1 or lipopolysaccharide (LPS). Activates IRF1 resulting in its rapid migration into the nucleus to mediate an efficientMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité