Anti-GPM6A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-10757R-A680
BOSSBS-10757R-A680EA 890 EUR InStock
BOSSBS-10757R-A680
Anti-GPM6A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Involved in neuronal differentiation, including differentiation and migration of neuronal stem cells. Plays a role in neuronal plasticity and is involved in neurite and filopodia outgrowth, filopodia motility and probably synapse formation. GPM6A-induced filopodia formation involves mitogen-activated protein kinase (MAPK) and Src Signalling pathways. May be involved in neuronal NGF-dependent Ca²⁺ influx. May be involved in regulation of endocytosis and intracellular trafficking of G-protein-coupled receptors (GPCRs); enhances internalisation and recycling of mu-type opioid receptor.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité