Anti-TNFRSF25 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-10938R-CY7
BOSSBS-10938R-CY7EA 900 EUR InStock
BOSSBS-10938R-CY7
Anti-TNFRSF25 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The protein encoded by this gene is a member of theTNF-receptor superfamily. This receptor is expressed preferentiallyin the tissues enriched in lymphocytes, and it may play a role inregulating lymphocyte homeostasis. This receptor has been shown tostimulate NF-kappa B activity and regulate cell apoptosis. Thesignal transduction of this receptor is mediated by various deathdomain containing adaptor proteins. Knockout studies in micesuggested the role of this gene in the removal of self-reactive Tcells in the thymus. Multiple alternatively spliced transcriptvariants of this gene encodingMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité