Anti-LSM14B Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-11001R-CY7
BOSSBS-11001R-CY7EA 900 EUR InStock
BOSSBS-11001R-CY7
Anti-LSM14B Rabbit Polyclonal Antibody (Cy7®)
Anticorps
Sm and Sm-like (LSm) proteins form donut-shaped, ubiquitously expressed heptameric complexes that are involved in various steps of RNA metabolism, including RNA-protein interactions and structural changes that are required during ribosomal subunit assembly. LSm14B, also known as C20orf40, FAM61B or LSM13, is a 385 amino acid protein that exists as multiple alternatively spliced isoforms and may play a role in RNA-related events. The gene encoding LSm14B maps to human chromosome 20. Comprising approximately 2% of the human genome, chromosome 20 contains nearly 63 million bases that encodeMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité