Anti-AKT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-11178R-A488
BOSSBS-11178R-A488EA 893 EUR InStock
BOSSBS-11178R-A488
Anti-AKT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
The endoplasmic reticulum exerts a quality control over newly synthesized proteins and a variety of components have been implicated in the specific recognition of aberrant or misfolded polypeptides. Signal peptide peptidase (SPP) catalyzes intramembrane proteolysis of some signal peptides after they have been cleaved from a preprotein, resulting in the release of the fragment from the ER membrane into the cytoplasm. SPP is required to generate lymphocyte cell surface (HLA-E) epitopes derived from MHC class I signal peptides, and may play a role in graft rejection. It also may be necessaryMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité