Anti-MYRF Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-11191R-A488
BOSSBS-11191R-A488EA 893 EUR InStock
BOSSBS-11191R-A488
Anti-MYRF Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Myelin regulatory factor: Consitutes a precursor of the transcription factor. Mediates the autocatalytic cleavage that releases the Myelin regulatory factor, N-terminal component that specifically activates transcription of central nervous system (CNS) myelin genes (PubMed:23966832). Myelin regulatory factor, C-terminal: Membrane-bound part that has no transcription factor activity and remains attached to the endoplasmic reticulum membrane following cleavage. Myelin regulatory factor, N-terminal: Transcription factor that specifically activates expression of myelin genes such as MBP, MOG,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité